N-Methyl-D-glucamine hydrochloride 97%

Synonyme: N-Methyl-D-glucamine Hydrochloride

BD145077-5G BD145077-1G BD145077-25G
BLDPBD145077-5GEA 56 EUR
BLDPBD145077-5G BLDPBD145077-1G BLDPBD145077-25G
N-Methyl-D-glucamine hydrochloride 97%
N-Methyl-D-glucamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₁₇NO₅
Molecular Weight: 231,67 g/mol
Lagertemperatur: Ambient
MDL: MFCD00060164
CAS-Nummer: 35564-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR