N-Methylacetamid 99% (durch GC)

Synonyme: N-methylacetamide

BD145046-25G
BLDPBD145046-25GEA 8 EUR
BLDPBD145046-25G
N-Methylacetamid 99% (durch GC)
N-Methylacetamid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃H₇NO
Molecular Weight: 73,09 g/mol
Lagertemperatur: Ambient
MDL: MFCD00008683
CAS-Nummer: 79-16-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR