N-Lauryldiethanolamine 99+%

Synonyme: N-Dodecyldiethanolamine,N,N-Bis(2-hydroxyethyl)dodecylamine,2,2'-(Dodecylazanediyl)diethanol

BD144932-25G BD144932-1000G BD144932-500G BD144932-100G
BLDPBD144932-25GEA 7 EUR
BLDPBD144932-25G BLDPBD144932-1000G BLDPBD144932-500G BLDPBD144932-100G
N-Lauryldiethanolamine 99+%
N-Lauryldiethanolamine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₃₅NO₂
Molecular Weight: 273,46 g/mol
Lagertemperatur: Ambient
MDL: MFCD00059002
CAS-Nummer: 1541-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR