N-Lauroyl-L-alanine 97%

BD144929-5G BD144929-25G BD144929-100G
BLDPBD144929-5GEA 21 EUR
BLDPBD144929-5G BLDPBD144929-25G BLDPBD144929-100G
N-Lauroyl-L-alanine 97%
N-Lauroyl-L-alanine

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₂₉NO₃
Molecular Weight: 271,39 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00216726
CAS-Nummer: 52558-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR