Nizatidine 98%

Synonyme: N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamineN-(2-(((2-((Dimethylamino)methyl)thiazol-4-yl)methyl)thio)ethyl)-N-methyl-2-nitroethene-1,1-diamine

BLDPBD144924-250MGEA 6 EUR
BLDPBD144924-250MG BLDPBD144924-1G
Nizatidine 98%
Nizatidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₂₁N₅O₂S₂
Molecular Weight: 331,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00865660
CAS-Nummer: 76963-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR