Nile blue chloride (Basic blue 12), Reagenzqualität

Synonyme: Nile Blue A chloride,5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium chloride

BD144763-5G BD144763-1G
BLDPBD144763-5GEA 31 EUR
BLDPBD144763-5G BLDPBD144763-1G
Nile blue chloride (Basic blue 12), Reagenzqualität
Nile blue chloride (Basic blue 12)
Formel: C₂₀H₂₀ClN₃O
Molecular Weight: 353,85 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011936
CAS-Nummer: 2381-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR