3-(4-Pyridyl)benzaldehyd 95%

Synonyme: 3-(Pyridin-4-yl)benzaldehyde,3-Pyrid-4-ylbenzaldehyde

BD14462-250MG BD14462-1G BD14462-5G
BLDPBD14462-250MGEA 21 EUR
BLDPBD14462-250MG BLDPBD14462-1G BLDPBD14462-5G
3-(4-Pyridyl)benzaldehyd 95%
3-(4-Pyridyl)benzaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₉NO
Molecular Weight: 183,2060 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD02684105
CAS-Nummer: 208190-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR