5-Amino-4-cyan-1-phenyl-1H-pyrazol-3-acetonitril 97%

Synonyme: 5-Amino-3-(cyanomethyl)-1-phenyl-1H-pyrazole-4-carbonitrile

BD14454-5G BD14454-25G BD14454-1G
BLDPBD14454-5GEA 66 EUR
BLDPBD14454-5G BLDPBD14454-25G BLDPBD14454-1G
5-Amino-4-cyan-1-phenyl-1H-pyrazol-3-acetonitril 97%
5-Amino-4-cyan-1-phenyl-1H-pyrazol-3-acetonitril

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₉N₅
Molecular Weight: 223,2334 g/mol
Lagertemperatur: Ambient
MDL: MFCD00020729
CAS-Nummer: 7152-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR