N-Methylheptan-1-amine 97%

BD144448-1G BD144448-10G BD144448-5G BD144448-25G BD144448-250MG
BLDPBD144448-1GEA 15 EUR
BLDPBD144448-1G BLDPBD144448-10G BLDPBD144448-5G BLDPBD144448-25G BLDPBD144448-250MG
N-Methylheptan-1-amine 97%
N-Methylheptan-1-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₉N
Molecular Weight: 129,24 g/mol
Lagertemperatur: Ambient
MDL: MFCD00039980
CAS-Nummer: 36343-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR