3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-bromophenyl)propanoic acid 98%

BLDPBD144367-250MGEA 45 EUR
BLDPBD144367-250MG BLDPBD144367-1G
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-bromophenyl)propanoic acid 98%
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-bromophenyl)propanoic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₄H₂₀BrNO₄
Molecular Weight: 466,32 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02090650
CAS-Nummer: 284492-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR