Neutralrot 98% BS
Lieferant: BLD PHARMATECH GMBH
|
|
Synonyme:
Basisrot 5
, 2-Amino-8-(dimethylamino)-3-methylphenaziniumchlorid
, N8,N8,3-Trimethyl-2,8-phenazindiamin Monohydrochlorid
BLDPBD144341-100GEA
33
EUR
BLDPBD144341-100G
BLDPBD144341-25G
BLDPBD144341-500G
Neutralrot 98% BS
Neutralrot
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₅H₁₇ClN₄ Molecular Weight: 288,78 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012651 CAS-Nummer: 553-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...