N-(3-Chlorobenzyl)ethanamine 97%

BD144277-1G
BLDPBD144277-1GEA 119 EUR
BLDPBD144277-1G
N-(3-Chlorobenzyl)ethanamine 97%
N-(3-Chlorobenzyl)ethanamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₂ClN
Molecular Weight: 169,65 g/mol
Lagertemperatur: Ambient
MDL: MFCD00045195
CAS-Nummer: 39180-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR