2-(Bromacetyl)naphthalin 95%

BLDPBD14422-25GEA 125 EUR
BLDPBD14422-25G BLDPBD14422-5G
2-(Bromacetyl)naphthalin 95%
2-(Bromacetyl)naphthalin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₉BrO
Molecular Weight: 249,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004109
CAS-Nummer: 613-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR