Nε-acetyl-L-lysine 97%

Synonyme: (S)-6-Acetamido-2-aminohexanoic acid,L-Lys(Ac)-OH,H-Lys(Ac)-OH,Nepsilon-acetyl-L-lysine

BD144183-10G BD144183-250MG BD144183-1G BD144183-5G BD144183-25G BD144183-100G
BLDPBD144183-10GEA 127 EUR
BLDPBD144183-10G BLDPBD144183-250MG BLDPBD144183-1G BLDPBD144183-5G BLDPBD144183-25G BLDPBD144183-100G
Nε-acetyl-L-lysine 97%
Nepsilon-acetyl-L-lysine

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₈H₁₆N₂O₃
Molecular Weight: 188,23 g/mol
Lagertemperatur: Freezer
MDL: MFCD00002639
CAS-Nummer: 692-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR