Neopentyl glycolbis(4-aminophenyl)ether 97%

Synonyme: 4,4'-((2,2-Dimethylpropane-1,3-diyl)bis(oxy))dianiline,2,2-Dimethylpropane 1,3-Bis(4-aminophenyl) Ether,1,3-Bis(4-aminophenoxy)neopentane,2,2-Bis[(4-aminophenoxy)methyl]propane

BD144154-5G BD144154-1G
BLDPBD144154-5GEA 32 EUR
BLDPBD144154-5G BLDPBD144154-1G
Neopentyl glycolbis(4-aminophenyl)ether 97%
Neopentyl glycolbis(4-aminophenyl)ether

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₇H₂₂N₂O₂
Molecular Weight: 286,38 g/mol
Lagertemperatur: Ambient
MDL: MFCD00671569
CAS-Nummer: 115570-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR