n-Dodecyl-β-D-maltosid 97%

Synonyme: Dodecyl-β-D-maltopyranosideLauryl-β-D-maltosideDDM

BLDPBD144059-1GEA 31 EUR
BLDPBD144059-1G BLDPBD144059-0.025 BLDPBD144059-250MG BLDPBD144059-10G BLDPBD144059-25G BLDPBD144059-5G
n-Dodecyl-β-D-maltosid 97%
n-Dodecyl-β-D-maltosid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₂₄H₄₆O₁₁
Molecular Weight: 510,6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00043012
CAS-Nummer: 69227-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR