Benzyl bis(2-chloroethyl)carbamate 95%

BD143900-5G BD143900-25G BD143900-10G BD143900-100G BD143900-1G
BLDPBD143900-5GEA 39 EUR
BLDPBD143900-5G BLDPBD143900-25G BLDPBD143900-10G BLDPBD143900-100G BLDPBD143900-1G
Benzyl bis(2-chloroethyl)carbamate 95%
Benzyl bis(2-chloroethyl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₅Cl₂NO₂
Molecular Weight: 276,16 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD06858694
CAS-Nummer: 72791-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR