(S)-1-Benzyl 2-methyl pyrrolidine-1,2-dicarboxylate 95%

BD143860-5G BD143860-25G BD143860-10G BD143860-500G BD143860-1G BD143860-100G
BLDPBD143860-5GEA 22 EUR
BLDPBD143860-5G BLDPBD143860-25G BLDPBD143860-10G BLDPBD143860-500G BLDPBD143860-1G BLDPBD143860-100G
(S)-1-Benzyl 2-methyl pyrrolidine-1,2-dicarboxylate 95%
(S)-1-Benzyl 2-methyl pyrrolidine-1,2-dicarboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₇NO₄
Molecular Weight: 263,29 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00134246
CAS-Nummer: 5211-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR