N-Butyl-2-chloroacetamide 97%

BLDPBD143758-250MGEA 15 EUR
BLDPBD143758-250MG BLDPBD143758-1G BLDPBD143758-5G
N-Butyl-2-chloroacetamide 97%
N-Butyl-2-chloroacetamide

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₁₂ClNO
Molecular Weight: 149,6200 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00776256
CAS-Nummer: 5349-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR