4-Brom-4'-fluorbiphenyl 95%

Synonyme: 1-Brom-4-(4-fluorphenyl)benzol

BD14373-100MG BD14373-5G BD14373-1G BD14373-250MG
BLDPBD14373-100MGEA 8 EUR
BLDPBD14373-100MG BLDPBD14373-5G BLDPBD14373-1G BLDPBD14373-250MG
4-Brom-4'-fluorbiphenyl 95%
4-Brom-4'-fluorbiphenyl

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₈BrF
Molecular Weight: 251,1 g/mol
Lagertemperatur: Ambient
MDL: MFCD00017954
CAS-Nummer: 398-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR