cis-N-Boc-4-cyano-L-proline methyl ester 98%

Synonyme: (2S,4S)-1-tert-Butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate,1-tert-Butyl 2-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate

BD143664-1G BD143664-5G BD143664-250MG
BLDPBD143664-1GEA 32 EUR
BLDPBD143664-1G BLDPBD143664-5G BLDPBD143664-250MG
cis-N-Boc-4-cyano-L-proline methyl ester 98%
cis-N-Boc-4-cyano-L-proline methyl ester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₈N₂O₄
Molecular Weight: 254,286 g/mol
Lagertemperatur: Ambient
MDL: MFCD03787928
CAS-Nummer: 487048-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR