Boc-4-methylaminobenzoic acid 95%

Synonyme: 4-((tert-Butoxycarbonyl)(methyl)amino)benzoic acid

BD143627-25G BD143627-250MG BD143627-1G BD143627-5G BD143627-100G
BLDPBD143627-25GEA 955 EUR
BLDPBD143627-25G BLDPBD143627-250MG BLDPBD143627-1G BLDPBD143627-5G BLDPBD143627-100G
Boc-4-methylaminobenzoic acid 95%
Boc-4-methylaminobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₇NO₄
Molecular Weight: 251,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD04972273
CAS-Nummer: 263021-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR