N-Benzyliminodiacetic acid 98%

BLDPBD143500-5GEA 164 EUR
BLDPBD143500-5G BLDPBD143500-1G
N-Benzyliminodiacetic acid 98%
N-Benzyliminodiacetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₃NO₄
Molecular Weight: 223,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004285
CAS-Nummer: 3987-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR