N-Benzylidenebenzenesulphonamide 98%

BD143495-250MG BD143495-1G BD143495-25G BD143495-5G
BLDPBD143495-250MGEA 15 EUR
BLDPBD143495-250MG BLDPBD143495-1G BLDPBD143495-25G BLDPBD143495-5G
N-Benzylidenebenzenesulphonamide 98%
N-Benzylidenebenzenesulphonamide

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₃H₁₁NO₂S
Lagertemperatur: Refrigerator
MDL: MFCD00011587
CAS-Nummer: 13909-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR