4-Nitro-1,8-naphthalsäureanhydrid 95%

Synonyme: 6-Nitrobenzo[de]isochromene-1,3-dione

BD14327-25G BD14327-1G BD14327-5G BD14327-10G BD14327-250MG
BLDPBD14327-25GEA 846 EUR
BLDPBD14327-25G BLDPBD14327-1G BLDPBD14327-5G BLDPBD14327-10G BLDPBD14327-250MG
4-Nitro-1,8-naphthalsäureanhydrid 95%
4-Nitro-1,8-naphthalsäureanhydrid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₅NO₅
Molecular Weight: 243,1718 g/mol
Lagertemperatur: Ambient
MDL: MFCD00013446
CAS-Nummer: 6642-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR