3-Nitro-1,8-naphthalsäureanhydrid 98%

Synonyme: 5-Nitrobenzo[de]isochromene-1,3-dione

BD14326-25G BD14326-1G BD14326-100G BD14326-5G
BLDPBD14326-25GEA 156 EUR
BLDPBD14326-25G BLDPBD14326-1G BLDPBD14326-100G BLDPBD14326-5G
3-Nitro-1,8-naphthalsäureanhydrid 98%
3-Nitro-1,8-naphthalsäureanhydrid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₅NO₅
Molecular Weight: 243,1718 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00006926
CAS-Nummer: 3027-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR