Dipentylsulfid 98%

Synonyme: Dipentylsulphane

BD143227-25G BD143227-5G
BLDPBD143227-25GEA 22 EUR
BLDPBD143227-25G BLDPBD143227-5G
Dipentylsulfid 98%
Dipentylsulfid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₂S
Molecular Weight: 174,35 g/mol
Lagertemperatur: Ambient
MDL: MFCD00027275
CAS-Nummer: 872-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR