(S)-N-(1-Amino-5-guanidino-1-oxopentan-2-yl)benzamide hydrochloride 95%

BD143168-250MG BD143168-1G BD143168-5G
BLDPBD143168-250MGEA 17 EUR
BLDPBD143168-250MG BLDPBD143168-1G BLDPBD143168-5G
(S)-N-(1-Amino-5-guanidino-1-oxopentan-2-yl)benzamide hydrochloride 95%
(S)-N-(1-Amino-5-guanidino-1-oxopentan-2-yl)benzamide hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₃H₂₀ClN₅O₂
Molecular Weight: 313,78 g/mol
Lagertemperatur: Ambient
MDL: MFCD00035003
CAS-Nummer: 4299-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR