Ac-DL-β-Phe-OH 97%

BLDPBD142986-1GEA 53 EUR
BLDPBD142986-1G BLDPBD142986-250MG
Ac-DL-β-Phe-OH 97%
Ac-DL-β-Phe-OH

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₃NO₃
Molecular Weight: 207,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00181164
CAS-Nummer: 40638-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR