N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid 97%

Synonyme: Boc-7-Ah,7-((tert-Butoxycarbonyl)amino)heptanoic acid,7-(tert-Butoxycarbonylamino)enanthic Acid,7-(Boc-amino)enanthic Acid,N-(tert-Butoxycarbonyl)-7-aminoenanthic Acid,7-(tert-Butoxycarbonylamino)heptanoic Acid,N-Boc-7-aminoheptanoic Acid,7-(Boc-amino)heptanoic Acid,N-Boc-7-aminoenanthic Acid

BD14297-10G BD14297-100G BD14297-5G BD14297-1G BD14297-25G BD14297-250MG
BLDPBD14297-10GEA 175 EUR
BLDPBD14297-10G BLDPBD14297-100G BLDPBD14297-5G BLDPBD14297-1G BLDPBD14297-25G BLDPBD14297-250MG
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid 97%
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₂₃NO₄
Molecular Weight: 245,32 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00063356
CAS-Nummer: 60142-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR