(S)-2-((S)-2-Acetamido-3-carboxypropanamido)pentanedioic acid

Lieferant: BLD PHARMATECH GMBH
BD142969-25MG
BLDPBD142969-25MGEA 105 EUR
BLDPBD142969-25MG
(S)-2-((S)-2-Acetamido-3-carboxypropanamido)pentanedioic acid
Proteine und Peptide
  • GHS07

Storage condition: Sealed in dry, Store in freezer, under -20°C
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR