5-Acetamido-2-hydroxybenzoic acid 98+%

Synonyme: 5-(Acetylamino)-2-hydroxybenzoic acid,5-Acetamidosalicylic acid

BD142953-250MG BD142953-1G BD142953-100MG BD142953-5G
BLDPBD142953-250MGEA 69 EUR
BLDPBD142953-250MG BLDPBD142953-1G BLDPBD142953-100MG BLDPBD142953-5G
5-Acetamido-2-hydroxybenzoic acid 98+%
5-Acetamido-2-hydroxybenzoic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₉NO₄
Molecular Weight: 195,172 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00463101
CAS-Nummer: 51-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR