Cyclohexylboronsäurepinakolester 98%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD14291-5GEA
31
EUR
BLDPBD14291-5G
BLDPBD14291-1G
BLDPBD14291-10G
BLDPBD14291-25G
BLDPBD14291-100G
Cyclohexylboronsäurepinakolester 98%
Cyclohexylboronsäurepinakolester
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₁₂H₂₃BO₂ Molecular Weight: 210,12 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04038749 CAS-Nummer: 87100-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...