1-Boc-isonipecotinsäuremethylester 98%

BLDPBD14276-5GEA 6 EUR
BLDPBD14276-5G BLDPBD14276-25G BLDPBD14276-10G BLDPBD14276-1000G BLDPBD14276-500G BLDPBD14276-100G
1-Boc-isonipecotinsäuremethylester 98%
1-Boc-isonipecotinsäuremethylester

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₂₁NO₄
Molecular Weight: 243,2994 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02183584
CAS-Nummer: 124443-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR