3-Amino-4-hydroxy-N,N-dimethylbenzenesulphonamide 98%

BD142683-250MG
BLDPBD142683-250MGEA 59 EUR
BLDPBD142683-250MG
3-Amino-4-hydroxy-N,N-dimethylbenzenesulphonamide 98%
3-Amino-4-hydroxy-N,N-dimethylbenzenesulphonamide

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₁₂N₂O₃S
Molecular Weight: 216,257 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00179913
CAS-Nummer: 24962-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR