1-Cyclohexenylboronsäurepinakolester 97%

Synonyme: 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,1-Cyclohexene-1-boronic acid pinacol ester

BD14267-250MG BD14267-5G BD14267-1G BD14267-10G BD14267-25G
BLDPBD14267-250MGEA 7 EUR
BLDPBD14267-250MG BLDPBD14267-5G BLDPBD14267-1G BLDPBD14267-10G BLDPBD14267-25G
1-Cyclohexenylboronsäurepinakolester 97%
1-Cyclohexenylboronsäurepinakolester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₂H₂₁BO₂
Molecular Weight: 208,1049 g/mol
Lagertemperatur: Freezer
MDL: MFCD05663845
CAS-Nummer: 141091-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR