2-Octylthiophen 97%

Synonyme: 2-n-Octylthiophene

BD14266-5G BD14266-1G
BLDPBD14266-5GEA 49 EUR
BLDPBD14266-5G BLDPBD14266-1G
2-Octylthiophen 97%
2-Octylthiophen

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₂H₂₀S
Molecular Weight: 196,36 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00041015
CAS-Nummer: 880-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR