2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose 97%
BLDPBD14262-500GEA 177 EUR
BLDPBD14262-500G BLDPBD14262-10G BLDPBD14262-5G BLDPBD14262-25G BLDPBD14262-100G
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose 97%
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose
Storage condition: Sealed in dry, Store in freezer, under -20°C
Formel: C₁₂H₂₀O₆
Molecular Weight: 260,29 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00022183
CAS-Nummer: 20880-92-6