Diethyl(4-methylbenzyl)phosphonat 97%

Synonyme: Diethyl 4-methylbenzylphosphonate

BD14252-100G BD14252-1G BD14252-5G BD14252-25G
BLDPBD14252-100GEA 378 EUR
BLDPBD14252-100G BLDPBD14252-1G BLDPBD14252-5G BLDPBD14252-25G
Diethyl(4-methylbenzyl)phosphonat 97%
Diethyl(4-methylbenzyl)phosphonat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₉O₃P
Molecular Weight: 242,26 g/mol
Lagertemperatur: Ambient
MDL: MFCD00041385
CAS-Nummer: 3762-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR