(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate 98%
Synonyme: 2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonicacidmonohydrate(-)-2,3:4,6-Di-O-isopropylidene-alpha-L-xylo-2-hexulofuranosonic acidDikegulac
BLDPBD14239-5GEA 15 EUR
BLDPBD14239-5G BLDPBD14239-25G BLDPBD14239-100G
(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate 98%
(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate
Storage condition: Sealed in dry, 2-8°C
Formel: C₁₂H₁₈O₇·H₂O
Molecular Weight: 274,27 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00150517
CAS-Nummer: 68539-16-2