N,N'-Diphenylbenzidin 98%

Synonyme: 4,4'-Bis(phenylamino)biphenyl,N4,N4'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine

BD142327-5G BD142327-100G BD142327-10G BD142327-1G BD142327-25G
BLDPBD142327-5GEA 18 EUR
BLDPBD142327-5G BLDPBD142327-100G BLDPBD142327-10G BLDPBD142327-1G BLDPBD142327-25G
N,N'-Diphenylbenzidin 98%
N,N'-Diphenylbenzidin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂₄H₂₀N₂
Molecular Weight: 336,44 g/mol
Lagertemperatur: Ambient
MDL: MFCD00003016
CAS-Nummer: 531-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR