1,3-Dibutylharnstoff 98%

Synonyme: N,N'-Dibutylharnstoff

BD142308-5G BD142308-100G BD142308-25G
BLDPBD142308-5GEA 24 EUR
BLDPBD142308-5G BLDPBD142308-100G BLDPBD142308-25G
1,3-Dibutylharnstoff 98%
1,3-Dibutylharnstoff

Storage condition: Sealed in dry, Room Temperature

Formel: H₃CCH₂CH₂CH₂NHC(O)NHCH₂CH₂CH₂CH₃
Molecular Weight: 172,27 g/mol
Lagertemperatur: Ambient
MDL: MFCD00015229
CAS-Nummer: 1792-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR