N,N-Dimethylschwefeldiamid 98%

Synonyme: N,N-Dimethylsulphamoylamide

BD142290-1G BD142290-5G BD142290-100G BD142290-10G BD142290-25G
BLDPBD142290-1GEA 8 EUR
BLDPBD142290-1G BLDPBD142290-5G BLDPBD142290-100G BLDPBD142290-10G BLDPBD142290-25G
N,N-Dimethylschwefeldiamid 98%
N,N-Dimethylsulphamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₂H₈N₂O₂S
Molecular Weight: 124,1600 g/mol
Lagertemperatur: Ambient
MDL: MFCD01861286
CAS-Nummer: 3984-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR