(Dimethylamino)(oxo)essigsäure 97%

BLDPBD142272-25GEA 38 EUR
BLDPBD142272-25G BLDPBD142272-500G BLDPBD142272-100G BLDPBD142272-5G BLDPBD142272-1G
(Dimethylamino)(oxo)essigsäure 97%
2-(Dimethylamino)-2-oxoacetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₄H₇NO₃
Molecular Weight: 117,1000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01971023
CAS-Nummer: 32833-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR