tert-Butyl-2-(aminomethyl)phenylcarbamate 97%

Synonyme: 2-(Aminomethyl)aniline, 1-BOC protected

BD14223-5G BD14223-250MG BD14223-1G BD14223-100MG
BLDPBD14223-5GEA 229 EUR
BLDPBD14223-5G BLDPBD14223-250MG BLDPBD14223-1G BLDPBD14223-100MG
tert-Butyl-2-(aminomethyl)phenylcarbamate 97%
tert-Butyl-2-(aminomethyl)phenylcarbamate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₂H₁₈N₂O₂
Molecular Weight: 222,29 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD04037902
CAS-Nummer: 849020-94-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR