N,N-Dimethyloctanamide 95%

Synonyme: N-Octanoyldimethylamine,N,N-Dimethylcaprylamide,Octanoic acid dimethylamide

BD142207-250MG BD142207-1G BD142207-5G
BLDPBD142207-250MGEA 10 EUR
BLDPBD142207-250MG BLDPBD142207-1G BLDPBD142207-5G
N,N-Dimethyloctanamide 95%
N,N-Dimethyloctanamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₁NO
Molecular Weight: 171,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD00043763
CAS-Nummer: 1118-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR