N,N-Dimethylbutyramide 98%

BD142205-5G BD142205-1G BD142205-25G BD142205-100G
BLDPBD142205-5GEA 40 EUR
BLDPBD142205-5G BLDPBD142205-1G BLDPBD142205-25G BLDPBD142205-100G
N,N-Dimethylbutyramide 98%
N,N-Dimethylbutyramide

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₁₃NO
Molecular Weight: 115,17 g/mol
Lagertemperatur: Ambient
MDL: MFCD00015222
CAS-Nummer: 760-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR