N,N-Dimethyl-4-fluor-2-nitroanilin 99%

Synonyme: 4-Fluoro-N,N-dimethyl-2-nitroaniline,N,N-Dimethyl 4-fluoro-2-nitroaniline

BD142180-1G BD142180-5G BD142180-100G BD142180-25G
BLDPBD142180-1GEA 7 EUR
BLDPBD142180-1G BLDPBD142180-5G BLDPBD142180-100G BLDPBD142180-25G
N,N-Dimethyl-4-fluor-2-nitroanilin 99%
N,N-Dimethyl-4-fluor-2-nitroanilin

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₉FN₂O₂
Molecular Weight: 184,17 g/mol
Lagertemperatur: Ambient
MDL: MFCD04116334
CAS-Nummer: 69261-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR