3,3'-Disulphanediylbis(N-methylpropanamide) 95%

BLDPBD142171-25GEA 77 EUR
BLDPBD142171-25G BLDPBD142171-1G BLDPBD142171-5G BLDPBD142171-100G
3,3'-Disulphanediylbis(N-methylpropanamide) 95%
3,3'-Disulphanediylbis(N-methylpropanamide)

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₁₆N₂O₂S₂
Molecular Weight: 236,35 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00172796
CAS-Nummer: 999-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR