2-Methoxy-N,N-dimethylacetamide 97%

BD142163-1G BD142163-25G BD142163-5G BD142163-100G
BLDPBD142163-1GEA 23 EUR
BLDPBD142163-1G BLDPBD142163-25G BLDPBD142163-5G BLDPBD142163-100G
2-Methoxy-N,N-dimethylacetamide 97%
2-Methoxy-N,N-dimethylacetamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₁₁NO₂
Molecular Weight: 117,14 g/mol
Lagertemperatur: Ambient
MDL: MFCD07787542
CAS-Nummer: 4128-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR